Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002147.2 | 1 - 2 | 126 | 75 | ||
GenBank mRNA | AK025690.1 | - | 1 - 2 | 126 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | karyopherin subunit alpha 7 | KPNA7 | IPOA8 | -54.0kb | Anti-sense | Chr. 7 - 99145467 - 99219417 on NCBI Build GRCh38 |
Downstream | actin related protein 2/3 complex subunit 1A | ARPC1A |
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L |
48.2kb | Sense | Chr. 7 - 99325873 - 99366262 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |