Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027989.hg.1 |
ENST00000266771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027989.hg.1 |
ENST00000544112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027989.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027989.hg.1 |
TC12002122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000535272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000535272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000366292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000376744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000535272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000535272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000539703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000545031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
ENST00000614634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012417.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197166.hg.1 |
TC1200012417.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000266771.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000376740.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
ENST00000544112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
NM_145648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197165.hg.1 |
TC1200012417.hg.1 |
Clariom D TC |