Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345.4 | NP_001336.2 | 6 - 7 | 660 | 75 | |
NM_201444.2 | NP_958852.1 | 6 - 7 | 705 | 75 | ||
NM_201445.1 | NP_958853.1 | 7 - 8 | 673 | 75 | ||
NM_201554.1 | NP_963848.1 | 6 - 7 | 573 | 75 | ||
XM_005268688.2 | XP_005268745.1 | 6 - 7 | 602 | 75 | ||
XM_011537991.2 | XP_011536293.1 | 7 - 8 | 770 | 75 | ||
XM_011537993.2 | XP_011536295.1 | 6 - 7 | 521 | 75 | ||
XM_017018909.1 | XP_016874398.1 | 5 - 6 | 491 | 75 | ||
GenBank mRNA | AF064769.1 | - | 6 - 7 | 582 | 75 | |
AK122973.1 | - | 4 - 5 | 2013 | 75 | ||
AK303320.1 | - | 6 - 7 | 598 | 75 | ||
AK307685.1 | - | 5 - 6 | 417 | 75 | ||
AK309939.1 | - | 6 - 7 | 549 | 75 | ||
AK310600.1 | - | 6 - 7 | 523 | 75 | ||
AY930112.1 | - | 6 - 7 | 600 | 75 | ||
BC023523.2 | - | 7 - 8 | 596 | 4304187 | 75 | |
BC031870.2 | - | 6 - 7 | 600 | 4819985 | 75 | |
BM910598.1 | - | 654 | 75 | |||
BQ943057.1 | - | 6 - 7 | 635 | 75 | ||
X62535.1 | - | 5 - 6 | 499 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |