Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242877.2 | NP_001229806.1 | 4 - 5 | 500 | 132 | |
NM_001242878.2 | NP_001229807.1 | 4 - 5 | 500 | 132 | ||
GenBank mRNA | AF332505.1 | - | 4 - 5 | 504 | 132 | |
AK022709.1 | - | 4 - 5 | 504 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |