Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018091.hg.1 |
ENST00000280871.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018091.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018091.hg.1 |
TC12001393.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018092.hg.1 |
ENST00000280871.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018092.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018092.hg.1 |
TC12001393.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178592.hg.1 |
TC1200010415.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000380858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000465517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
ENST00000505338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010415.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178593.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178593.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178593.hg.1 |
ENST00000280871.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178593.hg.1 |
NM_052885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178593.hg.1 |
TC1200010415.hg.1 |
Clariom D TC |