Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198695.2 | NP_941968.2 | 1 | 238 | 151 | |
GenBank mRNA | AB076355.1 | - | 1 | 238 | 151 | |
BC127016.1 | - | 1 | 232 | 40119338 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412919 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075297 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412917 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412909 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412914 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412903 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3293760 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30465 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3293760 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3293760 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075287 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075292 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412914 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412911 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412919 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075286 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412917 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412910 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412923 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075279 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412908 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412909 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412907 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075297 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412905 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412903 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075290 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075288 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412920 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412904 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412916 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412906 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412921 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412915 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412903 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412909 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075286 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412904 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412916 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412921 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412911 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412917 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412910 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412919 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412905 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412923 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412914 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075292 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412909 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412906 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075286 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412915 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075279 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075287 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075297 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412903 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412920 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412908 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075288 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11412907 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075294 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32075290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412903 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412909 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075286 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412925 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412915 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412921 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412923 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412917 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412924 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11412914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412905 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412903 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412915 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412948 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412912 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412917 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412923 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412921 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412919 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17242241 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412909 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412906 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_198695_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8069149 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8069148 | - | - |
Affymetrix Human Genome U219 Array | 11715321_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11715321_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR21002757.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002757.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002757.hg.1 | TC21000232.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | TC2100007375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149926.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | TC2100007375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149927.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149929.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149929.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149929.hg.1 | TC2100007375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100149929.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149929.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000334662.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000609664.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | ENST00000616689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198689.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007375.hg.1 | NM_198695.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019350 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019350 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 707754 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1793716 | BC127016.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_10900-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2000577 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_10900 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1793716 | AB076355.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1793716 | BC127016.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1793716 | NM_198695.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1793716 | AB076355.1 | human_transcriptome |