Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014824.hg.1 |
TC0X001088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014824.hg.1 |
ENST00000358697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014824.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014825.hg.1 |
TC0X001088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014825.hg.1 |
ENST00000358697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014825.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167404.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167404.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167404.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167404.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167404.hg.1 |
TC0X00009855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009855.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167406.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167406.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167406.hg.1 |
ENST00000358697.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167406.hg.1 |
NM_007156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167406.hg.1 |
TC0X00009855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
ENST00000374888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007424.hg.1 |
NM_007157.3 |
Refseq |