Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006898.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006898.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006898.hg.1 |
TC10000628.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008380.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156762.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156762.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156762.hg.1 |
ENST00000404190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156762.hg.1 |
NM_001080518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156762.hg.1 |
TC1000008380.hg.1 |
Clariom D TC |