Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005572.hg.1 |
ENST00000380612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005572.hg.1 |
ENST00000489821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005572.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005572.hg.1 |
TC21000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000329618.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380604.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008176.hg.1 |
NR_026543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
TC2100008176.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155304.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
TC2100008176.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155305.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
NR_026542.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
TC2100008176.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
ENST00000380612.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
ENST00000489821.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155306.hg.1 |
NR_026542.1 |
Refseq |