Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022454.hg.1 |
ENST00000300873.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022454.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022454.hg.1 |
TC19001651.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010996.hg.1 |
ENST00000624917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010995.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010995.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010995.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010995.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191512.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191512.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191512.hg.1 |
TC1900010995.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191512.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191512.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010995.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010995.hg.1 |
ENST00000300873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010995.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010995.hg.1 |
NM_033258.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010996.hg.1 |
ENST00000624917.1 |
Ensembl |