Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
variable charge, X-linked 3A |
VCX3A |
VCX-8r, VCX-A, VCX3, VCX8R, VCXA |
-527.8kb | Sense | Chr. X - 6533618 - 6535118 on NCBI Build GRCh38 |
Downstream |
steroid sulfatase (microsomal), isozyme S |
STS |
ARSC, ARSC1, ASC, ES, SSDD, XLI |
82.0kb | Anti-sense | Chr. X - 7147252 - 7354643 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |