Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fukutin | FKTN |
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4 |
-15.3kb | Sense | Chr. 9 - 105558117 - 105655950 on NCBI Build GRCh38 |
Downstream | TAL bHLH transcription factor 2 | TAL2 | 4.5kb | Sense | Chr. 9 - 105662457 - 105663112 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |