Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NFU1 iron-sulfur cluster scaffold | NFU1 |
CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU |
-67.5kb | Sense | Chr. 2 - 69396113 - 69438123 on NCBI Build GRCh38 |
Downstream |
small nucleolar RNA, H/ACA box 36C |
SNORA36C | 11.8kb | Sense | Chr. 2 - 69520044 - 69520171 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |