Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MT-RNR2-like 6 | MTRNR2L6 | HN6 | -43.2kb | Sense | Chr. 7 - 142666299 - 142667693 on NCBI Build GRCh38 |
Downstream | T cell receptor beta variable 27 | TRBV27 |
TCRBV14S1, TCRBV27S1 |
389.0 | Sense | Chr. 7 - 142715389 - 142715861 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |