Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928605 | LOC101928605 | Anti-sense | Chr. 7 - 144195321 - 144299589 on NCBI Build GRCh38 | ||
Downstream | olfactory receptor family 2 subfamily A member 7 | OR2A7 |
HSDJ0798C17, OR2A21 |
5.1kb | Sense | Chr. 7 - 144258696 - 144259628 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |