Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04399402_m1
See other ARNTL GE Assays ›
Gene Symbol:
ARNTL
Gene Name:
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases:
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location:
Chr.11: 13276613 - 13387268 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04399402_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ARNTL
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001030272.2
NP_001025443.1
5 - 6
580
93
NM_001030273.2
NP_001025444.1
6 - 7
651
93
NM_001178.5
NP_001169.3
6 - 7
635
93
NM_001297719.1
NP_001284648.1
6 - 7
635
93
NM_001297722.1
NP_001284651.1
5 - 6
580
93
NM_001297724.1
NP_001284653.1
6 - 7
644
93
XM_006718233.3
XP_006718296.1
5 - 6
522
93
XM_006718234.1
XP_006718297.1
6 - 7
588
93
XM_011520105.2
XP_011518407.2
4 - 5
686
93
XM_011520107.2
XP_011518409.2
4 - 5
681
93
XM_011520108.1
XP_011518410.1
6 - 7
628
93
XM_011520109.1
XP_011518411.1
4 - 5
383
93
XM_011520110.1
XP_011518412.1
7 - 8
643
93
XM_011520111.1
XP_011518413.1
5 - 6
458
93
XM_011520112.2
XP_011518414.1
7 - 8
643
93
XM_011520113.1
XP_011518415.1
7 - 8
643
93
XM_017017738.1
XP_016873227.1
4 - 5
684
93
XM_017017739.1
XP_016873228.1
4 - 5
680
93
XM_017017740.1
XP_016873229.1
5 - 6
522
93
XM_017017741.1
XP_016873230.1
4 - 5
383
93
XM_017017742.1
XP_016873231.1
4 - 5
383
93
XM_017017743.1
XP_016873232.1
4 - 5
383
93
XM_017017744.1
XP_016873233.1
7 - 8
643
93
XM_017017745.1
XP_016873234.1
6 - 7
588
93
XM_017017746.1
XP_016873235.1
6 - 7
598
93
XM_017017747.1
XP_016873236.1
5 - 6
458
93
XM_017017748.1
XP_016873237.1
5 - 6
515
93
GenBank mRNA
AB000812.1
-
3 - 4
216
93
AB000815.1
-
4 - 5
287
93
AB000816.1
-
5 - 6
627
93
AF044288.1
-
6 - 7
545
93
AK095749.1
-
6 - 7
536
93
AK291510.1
-
5 - 6
493
93
AK315174.1
-
6 - 7
573
93
BC016674.1
-
3 - 4
179
3921870
93
BC031214.1
-
6 - 7
476
4793007
93
BC041129.1
-
6 - 7
559
5753529
93
D89722.1
-
3 - 4
192
93
U60415.1
-
5 - 6
479
93
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ARNTL
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
circadian rhythm
negative regulation of TOR signaling
circadian regulation of gene expression
regulation of protein catabolic process
regulation of hair cycle
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of neurogenesis
regulation of insulin secretion
regulation of cell cycle
response to redox state
maternal process involved in parturition
positive regulation of canonical Wnt signaling pathway
oxidative stress-induced premature senescence
regulation of type B pancreatic cell development
negative regulation of glucocorticoid receptor signaling pathway
regulation of cellular senescence
positive regulation of skeletal muscle cell differentiation
transcription regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
core promoter sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
protein binding
aryl hydrocarbon receptor binding
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
Hsp90 protein binding
repressing transcription factor binding
E-box binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0