Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284113.1 | NP_001271042.1 | 2 - 3 | 353 | 107 | |
GenBank mRNA | AB168795.1 | - | 2 - 3 | 353 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |