Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568967.2 | XP_005569024.1 | 1 - 2 | 138 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |