Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283562.1 | NP_001270491.1 | 2 - 3 | 291 | 63 | |
GenBank mRNA | AB083313.2 | - | 2 - 3 | 291 | 63 | |
AB170659.1 | - | 2 - 3 | 262 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |