Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574854.2 | XP_005574911.1 | 2 - 3 | 982 | 65 | |
XM_005574855.2 | XP_005574912.1 | 2 - 3 | 1006 | 65 | ||
XM_005574856.2 | XP_005574913.1 | 2 - 3 | 794 | 65 | ||
XM_015433097.1 | XP_015288583.1 | 2 - 3 | 517 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |