Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579046.2 | XP_005579103.1 | 7 - 8 | 1452 | 72 | |
XM_005579047.2 | XP_005579104.1 | 7 - 8 | 1452 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |