Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588394.2 | XP_005588451.1 | 2 - 3 | 245 | 99 | |
GenBank mRNA | AB052143.1 | - | 2 - 3 | 275 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |