Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590471.2 | XP_005590528.1 | 4 - 5 | 612 | 97 | |
XM_015441561.1 | XP_015297047.1 | 5 - 6 | 628 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |