Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015443218.1 | XP_015298704.1 | 5 - 6 | 1556 | 98 | |
XM_015443219.1 | XP_015298705.1 | 5 - 6 | 1556 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |