Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102137003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577575.2 | XP_005577632.1 | 1 - 2 | 487 | 78 | |
GenBank mRNA | AB171556.1 | - | 1 - 2 | 355 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |