Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286147.1 | NP_001273076.1 | 6 - 7 | 796 | 105 | |
XM_015436506.1 | XP_015291992.1 | 7 - 8 | 882 | 105 | ||
XM_015436507.1 | XP_015291993.1 | 7 - 8 | 1043 | 105 | ||
GenBank mRNA | AB174101.1 | - | 6 - 7 | 796 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |