Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2U | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285106.1 | NP_001272035.1 | 4 - 5 | 756 | 115 | |
XM_015452091.1 | XP_015307577.1 | 4 - 5 | 559 | 115 | ||
GenBank mRNA | AB072767.1 | - | 4 - 5 | 756 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |