Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008300.3 | NP_032326.3 | 17 - 18 | 2479 | 88 | |
GenBank mRNA | AK133772.1 | - | 17 - 18 | 2422 | 88 | |
AK148883.1 | - | 6 - 7 | 754 | 88 | ||
AK155305.1 | - | 17 - 18 | 2396 | 88 | ||
AK166102.1 | - | 17 - 18 | 2462 | 88 | ||
AK220167.1 | - | 17 - 18 | 2430 | 88 | ||
AW545353.1 | - | 2 - 3 | 88 | |||
BC003770.1 | - | 17 - 18 | 2385 | 3494648 | 88 | |
BC003950.1 | - | 17 - 18 | 2372 | 88 | ||
D85904.1 | - | 17 - 18 | 2178 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |