Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009307.3 | NP_033333.2 | 3 - 4 | 563 | 79 | |
GenBank mRNA | AF257304.1 | - | 2 - 3 | 318 | 79 | |
AK036357.1 | - | 3 - 4 | 564 | 79 | ||
BC027019.1 | - | 2 - 3 | 324 | 5362352 | 79 | |
D37793.1 | - | 1 - 2 | 197 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |