Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010285.2 | NP_034415.1 | 2 - 3 | 178 | 84 | |
GenBank mRNA | AK005392.1 | - | 2 - 3 | 179 | 84 | |
AK018796.1 | - | 2 - 3 | 179 | 84 | ||
AK018816.1 | - | 2 - 3 | 179 | 84 | ||
AY902312.1 | - | 2 - 3 | 136 | 84 | ||
BC068204.1 | - | 2 - 3 | 155 | 1496474 | 84 | |
M31658.1 | - | 2 - 3 | 165 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |