Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011863.2 | NP_035993.1 | 1 - 2 | 212 | 65 | |
GenBank mRNA | AK146507.1 | - | 1 - 2 | 85 | 65 | |
AK153296.1 | - | 1 - 2 | 82 | 65 | ||
AK168982.1 | - | 1 - 2 | 85 | 65 | ||
AK169124.1 | - | 1 - 2 | 79 | 65 | ||
U34883.1 | - | 1 - 2 | 89 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |