Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159509.1 | NP_001152981.1 | 4 - 5 | 674 | 53 | |
NM_001159510.1 | NP_001152982.1 | 4 - 5 | 625 | 53 | ||
NM_011862.3 | NP_035992.1 | 4 - 5 | 713 | 53 | ||
GenBank mRNA | AF128535.1 | - | 4 - 5 | 703 | 53 | |
AK154785.1 | - | 4 - 5 | 600 | 53 | ||
AK170299.1 | - | 4 - 5 | 672 | 53 | ||
BC023502.1 | - | 5 - 6 | 579 | 5031043 | 53 | |
CA560112.1 | - | 2 - 3 | 358 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |