Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013781.3 | NP_038809.1 | 3 - 4 | 645 | 93 | |
GenBank mRNA | AB043953.1 | - | 3 - 4 | 628 | 93 | |
AF168364.1 | - | 3 - 4 | 586 | 93 | ||
AK155165.1 | - | 3 - 4 | 581 | 93 | ||
BC113203.1 | - | 3 - 4 | 600 | 40048064 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |