Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018822.3 | NP_061292.2 | 3 - 4 | 382 | 66 | |
GenBank mRNA | AF153827.1 | - | 3 - 4 | 356 | 66 | |
AF304053.1 | - | 3 - 4 | 356 | 66 | ||
AF304054.1 | - | 3 - 4 | 356 | 66 | ||
AK034220.1 | - | 3 - 4 | 293 | 66 | ||
AK052266.1 | - | 3 - 4 | 375 | 66 | ||
AK076267.1 | - | 3 - 4 | 411 | 66 | ||
AK141266.1 | - | 3 - 4 | 330 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |