Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031408.2 | NP_113596.2 | 4 - 5 | 446 | 134 | |
GenBank mRNA | AY176042.1 | - | 4 - 5 | 462 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |