Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147201.2 | NP_671734.1 | 10 - 11 | 1001 | 65 | |
GenBank mRNA | AF302138.1 | - | 10 - 11 | 1000 | 65 | |
AK046142.1 | - | 11 - 12 | 1019 | 65 | ||
AK077440.1 | - | 10 - 11 | 1037 | 65 | ||
AK170501.1 | - | 10 - 11 | 1017 | 65 | ||
AK188331.1 | - | 2 - 3 | 142 | 65 | ||
AK207083.1 | - | 2 - 3 | 142 | 65 | ||
BC004756.1 | - | 10 - 11 | 971 | 3154089 | 65 | |
BC018463.1 | - | 10 - 11 | 975 | 3598438 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |