Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026313.1 | NP_080589.1 | 9 - 10 | 1260 | 90 | |
GenBank mRNA | AK084714.1 | - | 4 - 5 | 647 | 90 | |
AK151228.1 | - | 9 - 10 | 1308 | 90 | ||
AK151839.1 | - | 9 - 10 | 1308 | 90 | ||
BC009092.1 | - | 9 - 10 | 1260 | 3157532 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |