Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033572.2 | NP_291050.1 | 4 - 5 | 690 | 77 | |
GenBank mRNA | AF410456.1 | - | 4 - 5 | 669 | 77 | |
AK005040.1 | - | 4 - 5 | 636 | 77 | ||
AK017733.1 | - | 4 - 5 | 689 | 77 | ||
BC024714.1 | - | 4 - 5 | 662 | 5134834 | 77 | |
BC055335.1 | - | 4 - 5 | 688 | 6412407 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |