Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039533.2 | NP_001034622.1 | 14 - 15 | 1415 | 87 | |
NM_001291017.1 | NP_001277946.1 | 14 - 15 | 1415 | 87 | ||
NM_053181.3 | NP_444411.2 | 14 - 15 | 1415 | 87 | ||
GenBank mRNA | AK004611.1 | - | 14 - 15 | 1292 | 87 | |
AK030613.1 | - | 14 - 15 | 1246 | 87 | ||
AK033107.1 | - | 14 - 15 | 1246 | 87 | ||
AK054329.1 | - | 14 - 15 | 1398 | 87 | ||
AK075829.1 | - | 7 - 8 | 538 | 87 | ||
AK163696.1 | - | 14 - 15 | 1286 | 87 | ||
AK165587.1 | - | 14 - 15 | 1250 | 87 | ||
BC005541.1 | - | 14 - 15 | 1233 | 3590316 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |