Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190437.1 | NP_001177366.1 | 3 - 4 | 375 | 70 | |
NM_145215.2 | NP_660250.1 | 3 - 4 | 852 | 70 | ||
NR_033794.1 | 2 - 3 | 837 | 70 | |||
GenBank mRNA | AF412033.1 | - | 3 - 4 | 852 | 70 | |
AK004244.1 | - | 3 - 4 | 386 | 70 | ||
BC038052.1 | - | 2 - 3 | 254 | 70 | ||
BC051486.1 | - | 3 - 4 | 70 | |||
BC056624.1 | - | 2 - 3 | 837 | 70 | ||
BC069866.1 | - | 3 - 4 | 474 | 6478980 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |