Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023596.3 | NP_076085.1 | 1 - 2 | 52 | 79 | |
GenBank mRNA | AF326986.1 | - | 1 - 2 | 63 | 79 | |
AK034133.1 | - | 1 - 2 | 48 | 79 | ||
AK051317.1 | - | 1 - 2 | 54 | 79 | ||
AK157868.1 | - | 1 - 2 | 53 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus 1700 Inventoried Probe spans exons |