Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025812.2 | NP_080088.1 | 6 - 7 | 697 | 126 | |
GenBank mRNA | AK004596.1 | - | 6 - 7 | 717 | 126 | |
AK017716.1 | - | 6 - 7 | 900 | 126 | ||
AK039222.1 | - | 6 - 7 | 719 | 126 | ||
AK129010.1 | - | 6 - 7 | 899 | 126 | ||
AK134213.1 | - | 6 - 7 | 869 | 126 | ||
BC013804.1 | - | 6 - 7 | 697 | 3602239 | 126 | |
BC068257.1 | - | 6 - 7 | 770 | 6434284 | 126 | |
DQ182735.1 | - | 6 - 7 | 697 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |