Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031251.4 | NP_112541.1 | 2 - 3 | 184 | 87 | |
GenBank mRNA | AJ250670.1 | - | 1 - 2 | 82 | 87 | |
AK054467.1 | - | 3 - 4 | 362 | 87 | ||
AK078511.1 | - | 2 - 3 | 196 | 87 | ||
AK079295.1 | - | 2 - 3 | 185 | 87 | ||
AK085517.1 | - | 3 - 4 | 361 | 87 | ||
AK137143.1 | - | 2 - 3 | 227 | 87 | ||
AK170790.1 | - | 2 - 3 | 196 | 87 | ||
AK170828.1 | - | 2 - 3 | 207 | 87 | ||
BC005479.1 | - | 2 - 3 | 171 | 3484952 | 87 | |
BC006033.1 | - | 2 - 3 | 171 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |