Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019993.3 | NP_064377.2 | 1 - 2 | 428 | 75 | |
GenBank mRNA | AB296386.1 | - | 1 - 2 | 373 | 75 | |
AF170919.1 | - | 1 - 2 | 173 | 75 | ||
AK132336.1 | - | 1 - 2 | 213 | 75 | ||
AK149589.1 | - | 1 - 2 | 214 | 75 | ||
AK150093.1 | - | 1 - 2 | 171 | 75 | ||
AK154914.1 | - | 1 - 2 | 412 | 75 | ||
AK162354.1 | - | 1 - 2 | 212 | 75 | ||
AK165730.1 | - | 1 - 2 | 428 | 75 | ||
AK168261.1 | - | 1 - 2 | 406 | 75 | ||
AK168309.1 | - | 1 - 2 | 406 | 75 | ||
AK168878.1 | - | 1 - 2 | 377 | 75 | ||
BC003297.1 | - | 1 - 2 | 205 | 3583976 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |