Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nosip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163684.1 | NP_001157156.1 | 2 - 3 | 183 | 66 | |
NM_025533.3 | NP_079809.1 | 2 - 3 | 183 | 66 | ||
GenBank mRNA | AK007962.1 | - | 2 - 3 | 132 | 66 | |
AK010006.1 | - | 2 - 3 | 119 | 66 | ||
BC089029.1 | - | 2 - 3 | 109 | 30624825 | 66 | |
DV648837.1 | - | 2 - 3 | 183 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |