Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008564.2 | NP_032590.2 | 4 - 5 | 808 | 70 | |
GenBank mRNA | AF004105.1 | - | 3 - 4 | 698 | 70 | |
AK037751.1 | - | 4 - 5 | 733 | 70 | ||
AK077801.1 | - | 4 - 5 | 712 | 70 | ||
AK088156.1 | - | 4 - 5 | 721 | 70 | ||
AK088378.1 | - | 4 - 5 | 69 | |||
AK129039.1 | - | 4 - 5 | 689 | 70 | ||
AK145176.1 | - | 4 - 5 | 719 | 70 | ||
AK146187.1 | - | 4 - 5 | 723 | 70 | ||
AK146375.1 | - | 4 - 5 | 732 | 70 | ||
BC055318.1 | - | 4 - 5 | 717 | 5696348 | 70 | |
D86725.1 | - | 4 - 5 | 728 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |