Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010247.2 | NP_034377.2 | 5 - 6 | 704 | 71 | |
GenBank mRNA | AB010282.1 | - | 5 - 6 | 645 | 71 | |
AF483500.1 | - | 4 - 5 | 584 | 71 | ||
AF483501.1 | - | 4 - 5 | 584 | 71 | ||
AK143570.1 | - | 5 - 6 | 679 | 71 | ||
AK146392.1 | - | 5 - 6 | 623 | 71 | ||
AK146394.1 | - | 5 - 6 | 623 | 71 | ||
AK146396.1 | - | 5 - 6 | 671 | 71 | ||
BC031422.1 | - | 5 - 6 | 653 | 3669715 | 71 | |
BC051085.1 | - | 5 - 6 | 1882 | 71 | ||
M38700.1 | - | 4 - 5 | 546 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |