Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013672.2 | NP_038700.2 | 4 - 5 | 1940 | 75 | |
GenBank mRNA | AF022363.1 | - | 4 - 5 | 1937 | 75 | |
AF062566.1 | - | 4 - 5 | 1876 | 75 | ||
CF729889.1 | - | 2 - 3 | 532 | 75 | ||
S79832.1 | - | 4 - 5 | 989 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus Mouse Hypoxia Panel Inventoried Probe spans exons |