Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019675.2 | NP_062649.1 | 2 - 3 | 127 | 76 | |
GenBank mRNA | AF105222.1 | - | 1 - 2 | 99 | 76 | |
BC014707.1 | - | 2 - 3 | 129 | 4507015 | 76 | |
BC046752.1 | - | 2 - 3 | 127 | 5693069 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |