Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271599.1 | NP_001258528.1 | 6 - 7 | 854 | 90 | |
NM_019972.3 | NP_064356.2 | 6 - 7 | 854 | 90 | ||
GenBank mRNA | AF175279.1 | - | 6 - 7 | 783 | 90 | |
AK147442.1 | - | 6 - 7 | 837 | 90 | ||
BC034129.1 | - | 3 - 4 | 287 | 90 | ||
BC056343.1 | - | 6 - 7 | 786 | 5704346 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |